missing translation for 'onlineSavingsMsg'
Learn More

Gibberellin A34 (>90%), TRC

Product Code. 17952215
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952215 0.5 mg 0.5mg
17920875 2.5 mg 2.5mg
17942775 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17952215 Supplier Toronto Research Chemicals Supplier No. TRCG3773700.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32630-92-5
Molecular Formula C19 H24 O6
Molecular Weight (g/mol) 348.39
Synonym GA34, 1,4a-Lactone-2β,3β,4a-trihydroxy-1-methyl-8-methylene-4aα,4bβ-gibbane-1α,10β-dicarboxylic Acid, 4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene-gibbane-1,10-dicarboxylic Acid Derivative;
SMILES C[C@]12[C@@H](O)[C@@H](O)C[C@@]3(OC1=O)[C@@H]4CC[C@@H]5C[C@]4(CC5=C)[C@H]([C@H]23)C(=O)O

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C19H24O6/c1-8-5-18-6-9(8)3-4-11(18)19-7-10(20)14(21)17(2,16(24)25-19)13(19)12(18)15(22)23/h9-14,20-21H,1,3-7H2,2H3,(H,22,23)/t9-,10+,11-,12-,13-,14+,17+,18+,19-/m1/s1
Formula Weight 348.1573
Chemical Name or Material Gibberellin A34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.