missing translation for 'onlineSavingsMsg'
Learn More

Gibberellic Acid Methyl Ester, TRC

Product Code. 30400332
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400332 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400332 Supplier Toronto Research Chemicals Supplier No. TRCG377310250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gibberellic Acid Methyl Ester
CAS 510-50-9
Molecular Formula C20H24O6
Quantity 250 mg
Synonym GA3 Methyl Ester, Gibberellin A3 Methyl Ester, Methyl Gibberellate, 4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene-gibb-3-ene-1,10-dicarboxylic Acid Derivative
InChI Formula InChI=1S/C20H24O6/c1-10-8-18-9-19(10,24)6-4-11(18)20-7-5-12(21)17(2,16(23)26-20)14(20)13(18)15(22)25-3/h5,7,11-14,21,24H,1,4,6,8-9H2,2-3H3/t11-,12+,13-,14-,17-,18+,19+,20-/m1/s1
SMILES COC(=O)[C@H]1[C@@H]2[C@@]3(C)[C@@H](O)C=C[C@@]2(OC3=O)[C@@H]4CC[C@]5(O)C[C@]14CC5=C
Molecular Weight (g/mol) 360.4
Formula Weight 360.1573
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.