missing translation for 'onlineSavingsMsg'
Learn More

Gibberellic acid, 99%

Product Code. 10702464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10702464 25 g Glass bottle 25g
10655491 1 g Glass bottle 1g
10219890 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
Product Code. 10702464 Supplier Thermo Scientific Acros Supplier No. 410910250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.

CAS: 77-06-5 | C19H22O6 | 346.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-06-5
Molecular Formula C19H22O6
Molecular Weight (g/mol) 346.38
MDL Number MFCD00079329
InChI Key IXORZMNAPKEEDV-QTWFBFKQSA-N
Synonym gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs
PubChem CID 91757643
IUPAC Name (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
SMILES CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O

Specifications

Melting Point 227°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 4454
Specific Rotation Condition 82.50 (20.00°C c=10,ethanol)
Specific Rotation 82.5
Solubility Information Solubility in water: 5g/L (20° c). Other solubilities: soluble in aq.sol. of sodium bicarbonate,soluble in methanol,ethanol,acetone and aq. sol.,of sodium acetate,moderately soluble in ethyl acetate- slightly,soluble in ether- insoluble in chl
Formula Weight 346.38
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Gibberellic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.