missing translation for 'onlineSavingsMsg'
Learn More

Gibberellic Acid-13C,d2 (90%), TRC

Product Code. 30395961
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395961 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30395961 Supplier Toronto Research Chemicals Supplier No. TRCG3774535MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gibberellic Acid 13C D2 (17-13C 17,17-D2)
CAS 120396-36-3
Purity Grade Notes HPLC
Molecular Formula 13C C18 D2 H20 O6
Quantity 5 mg
Synonym Gibberellic Acid-13C,d2, Gibberellic Acid 13C D2 (17-13C 17,17-D2)
InChI Formula InChI=1S/C19H22O6/c1-9-7-17-8-18(9,24)5-3-10(17)19-6-4-11(20)16(2,15(23)25-19)13(19)12(17)14(21)22/h4,6,10-13,20,24H,1,3,5,7-8H2,2H3,(H,21,22)/t10-,11+,12-,13-,16-,17+,18+,19-/m1/s1/i1+1D2
SMILES [2H][13C](=C1C[C@]23C[C@@]1(O)CC[C@H]2[C@]45OC(=O)[C@](C)([C@@H](O)C=C4)[C@H]5[C@@H]3C(=O)O)[2H]
Molecular Weight (g/mol) 349.379
Formula Weight 349.158
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.