missing translation for 'onlineSavingsMsg'
Learn More

Giardia lamblia Mouse anti-Protozoa, Clone: BB1.1E5, Novus Biologicals™

Product Code. 18380783 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380783 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380783 Supplier Novus Biologicals Supplier No. NBP30754620UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Giardia lamblia Monoclonal specifically detects Giardia lamblia in Protozoa samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Giardia lamblia
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone BB1.1E5
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias G lamblia; Giardia intestinalis
Host Species Mouse
Immunogen Giardia lamblia
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.