missing translation for 'onlineSavingsMsg'
Learn More

Giardia lamblia Antibody (BB1.1E5), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18576506 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18576506 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18576506 Supplier Novus Biologicals Supplier No. NBP308870AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Giardia lamblia Monoclonal antibody specifically detects Giardia lamblia in Protozoa samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Giardia lamblia
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone BB1.1E5
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias G lamblia; Giardia intestinalis
Host Species Mouse
Immunogen Giardia lamblia
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.