Learn More
Invitrogen™ Giardia lamblia 65 kDa Monoclonal Antibody (6291)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547567
Description
Specificity: Specific to the 65 kDa Giardia antigen. Detects cysts. Comments: Best ELISA pairs (capture/conjugate): Giardia lamblia 65 kDa Monoclonal antibody (Product # MA5-47567)/Giardia lamblia 65 kDa Monoclonal antibody (Product # MA5-47568); Giardia lamblia 65 kDa Monoclonal antibody (Product # MA5-47567)/Giardia lamblia 65 kDa Polyclonal antibody (Product # MA1-7437); Giardia lamblia 65 kDa Monoclonal antibody (Product # MA5-47567)/Giardia lamblia 65 kDa Polyclonal antibody (Product # MA1-7440). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Giardiasisis a diarrhoeal illness caused by a single celled microscopic protozoan parasite, Giardia lamblia, also known as Giardia intestinalis. Giardia lamblia exists in two forms, an active form called a trophozoite, and an inactive form called a cyst. The active trophozoite attaches to the lining of the small intestine and is responsible for causing the signs and symptoms of giardiasis. The trophozoite cannot live long outside of the body and spread of infection is via the cyst which is excreted in the host's faeces. When it is ingested, stomach acid activates the cyst, and the cyst develops into the disease causing trophozoite in the new host. Giardiasis is diagnosed by finding cysts or trophozoites in the faeces.
Specifications
| Giardia lamblia 65 kDa | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Protozoa | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| 6291 | |
| Unconjugated | |
| G. lamblia | |
| Proprietary sequence. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.