missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GI24 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24849-1-AP-150UL
This item is not returnable.
View return policy
Description
VISTA is a homolog of PD-1 (PD-1H). PD-1H is a 309 aa type I transmembrane protein that belongs to the CD28 superfamily. This cell surface receptor is expressed on nearly all hematopoietic cells except B cells. VISTA negatively regulates T cell responses by suppressing early TCR activation, inhibiting cell proliferation, and suppressing TH1 cytokine production.Specifications
| GI24 | |
| Polyclonal | |
| Unconjugated | |
| VSIR | |
| B7-H5, C10orf54, GI24, Platelet receptor Gi24, PP2135, SISP1, UNQ730/PRO1412, VISTA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64115 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9H7M9 | |
| VSIR | |
| GI24 Fusion Protein Ag20600 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction