missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Limpet Labels GHS Health Hazard Labels, 20 x 20mm
Description
- Hazard warning diamonds fully comply with the GHS, CLP, RID IMDG/IMO, ADN, & ICAO/IATA requirements
- Excellent weather, chemical, and abrasion resistance
- Marine grade adhesive
- 1000 labels per roll
Specifications
Specifications
| Certifications/Compliance | GHS, CLP, RID IMDG/IMO, ADN, & ICAO/IATA requirements |
| Finish | Laminated |
| Material | Polypropylene |
| Quantity | 1000/Roll |
| Resistant To | Weather, chemicals, abrasion |
| Description | PP (Polypropylene) |
| Mounting Type | Self-adhesive |
| Height (Metric) | 20 mm |
| Width (Metric) | 20 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?