missing translation for 'onlineSavingsMsg'
Learn More

GHRP 3 TFA Salt, TRC

Product Code. 30393900
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393900 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393900 Supplier Toronto Research Chemicals Supplier No. TRCG93212510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material GHRP 3 TFA Salt
CAS 259230-56-3
Molecular Formula C32H50N10O5 . xC2HF3O2
Quantity 10 mg
Synonym 2-Methylalanyl-D-tryptophyl-D-prolyl-D-isoleucyl-L-argininamide, Human growth hormone-releasing peptide 3 TFA Salt
InChI Formula InChI=1S/C32H50N10O5/c1-5-18(2)25(28(45)39-22(26(33)43)12-8-14-37-31(34)35)41-27(44)24-13-9-15-42(24)29(46)23(40-30(47)32(3,4)36)16-19-17-38-21-11-7-6-10-20(19)21/h6-7,10-11,17-18,22-25,38H,5,8-9,12-16,36H2,1-4H3,(H2,33,43)(H,39,45)(H,40,47)(H,41,44)(H4,34,35,37)/t18-,22+,23-,24-,25-/m1/s1
SMILES CC[C@@H](C)[C@@H](NC(=O)[C@H]1CCCN1C(=O)[C@@H](Cc2c[nH]c3ccccc23)NC(=O)C(C)(C)N)C(=O)N[C@@H](CCCNC(=N)N)C(=O)N
IUPAC Name (2R)-N-[(2R,3R)-1-[[(2S)-1-amino-5-carbamimidamido-1-oxopentan-2-yl]amino]-3-methyl-1-oxopentan-2-yl]-1-[(2R)-2-[(2-amino-2-methylpropanoyl)amino]-3-(1H-indol-3-yl)propanoyl]pyrrolidine-2-carboxamide
Molecular Weight (g/mol) 654.8
Formula Weight 654.3966
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.