missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GHRHR Monoclonal Antibody (1037442)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA560906

Product Code. 30535127

  • £324.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Growth Hormone Releasing Hormone (GHRH) is a member of a family of peptide hormones that also includes VIP (vasoactive intestinal polypeptide) and PACAP (pituitary adenylate cyclase-activating polypeptide). It is a hypothalamic peptide that binds to G-protein coupled receptors to stimulate secretion and synthesis of growth hormone (GH). GHRH receptor proteins have been identified on the plasma membrane of primary human tumor cells, and are thought to be involved in the growth of a variety of cancer cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GHRHR
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q02643
Ghrhr
Human GHRHR synthetic peptide.
100 μg
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Immunocytochemistry
1037442
Unconjugated
Ghrhr
GHRFR; GHRH receptor; GHRHR; Ghrh-r; GHRHREC; GHRHRpsv; GRF receptor; GRFR; Growth hormone - releasing receptor; growth hormone releasing hormone receptor; growth hormone-releasing factor receptor; Growth hormone-releasing hormone receptor; IGHD1B; lit; little
Mouse
Protein A/G
RUO
2692
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.