missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GHRH Monoclonal Antibody (C2), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543802
This item is not returnable.
View return policy
Description
The activated kinase acts on a variety of targets. Likely to be the GTPase effector that links the Rho-related GTPases to the JNK MAP kinase pathway. Activated by CDC42 and RAC1. Involved in dissolution of stress fibers and reorganization of focal complexes. Involved in regulation of microtubule biogenesis through phosphorylation of TBCB. Activity is inhibited in cells undergoing apoptosis, potentially due to binding of CDC2L1 and CDC2L2.
Specifications
| GHRH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P01286 | |
| Ghrh | |
| Cys19-Gly108 (Accession P01286), with N-terminal His and GST Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C2 | |
| Unconjugated | |
| Ghrh | |
| GHRF; Ghrh; GRF; growth hormone releasing hormone; growth hormone-releasing factor; Growth hormone-releasing hormone; INN; MGC119781; OTTHUMP00000030915; Sermorelin; somatocrinin; Somatoliberin; somatorelin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2691 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction