missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ghrelin Receptor/GHSR Antibody (502430), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8370B
This item is not returnable.
View return policy
Description
Ghrelin Receptor/GHSR Monoclonal antibody specifically detects Ghrelin Receptor/GHSR in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Ghrelin Receptor/GHSR | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 502430 | |
| Flow Cytometry, CyTOF-ready | |
| GH-releasing peptide receptor, Ghrelin receptor, GHRP, GHS-R, growth hormone secretagogue receptor, growth hormone secretagogue receptor type 1 | |
| NS0 mouse myeloma cell line transfected with human GHSR, Accession # NP_940799 | |
| 0.1 mL | |
| GPCR | |
| 2693 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction