missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ghrelin/Obestatin Antibody (45), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47183MFV610
This item is not returnable.
View return policy
Description
Ghrelin/Obestatin Monoclonal antibody specifically detects Ghrelin/Obestatin in Human samples. It is validated for Western Blot, ELISA
Specifications
| Ghrelin/Obestatin | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 45 | |
| Western Blot, ELISA | |
| appetite-regulating hormone, ghrelin, ghrelin/obestatin preprohormone, ghrelin/obestatin prepropeptide, Growth hormone secretagogue, Growth hormone-releasing peptide, Motilin-related peptide, MTLRPgrowth hormone secretagogue receptor ligand, obestatin, Protein M46 | |
| Synthetic human Ser3-octanoyl ghrelin-28 | |
| 0.1 mL | |
| Angiogenesis, Neuroscience | |
| 51738 | |
| Store at 4C in the dark. | |
| IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction