missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GHITM Polyclonal specifically detects GHITM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | GHITM |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Dermal papilla-derived protein 2, DERP2MICS1, DKFZp566C0746, FLJ26584, growth hormone inducible transmembrane protein, growth hormone-inducible transmembrane protein, HSPC282, Mitochondrial morphology and cristae structure 1, PTD010, TMBIM5My021, transmembrane BAX inhibitor motif containing 5, Transmembrane BAX inhibitor motif-containing protein 5 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human GHITM fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?