missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GHITM Antibody (42), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-12773AF405
This item is not returnable.
View return policy
Description
GHITM Monoclonal antibody specifically detects GHITM in Invertebrate samples. It is validated for ELISA
Specifications
| GHITM | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Dermal papilla-derived protein 2, DERP2MICS1, DKFZp566C0746, FLJ26584, growth hormone inducible transmembrane protein, growth hormone-inducible transmembrane protein, HSPC282, Mitochondrial morphology and cristae structure 1, PTD010, TMBIM5My021, transmembrane BAX inhibitor motif containing 5, Transmembrane BAX inhibitor motif-containing protein 5 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dermatophagoides pteronyssinus GHITM (Uniprot#: P49278; Gln19-Asp146). | |
| 100 μg | |
| Primary | |
| Invertebrate | |
| Purified |
| ELISA | |
| 42 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 27069 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction