missing translation for 'onlineSavingsMsg'
Learn More

GGTI298 Trifluoroacetate, MedChemExpress

Product Code. 16443506
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443506

Brand: Medchem Express HY15871

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GGTI298 Trifluoroacetate is a CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor, which can inhibit Rap1A with IC50 of 3 μM; little effect on Ha-Ras with IC50 of >20 μM.

GGTI298 Trifluoroacetate is a CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor, which can inhibit Rap1A with IC50 of 3 μM

Specifications

Chemical Name or Material GGTI298 Trifluoroacetate
CAS 1217457-86-7
Color White
Purity Grade Notes Research
Molecular Formula C29H34F3N3O5S
Quantity 25 mg
Solubility Information DMSO : 150 mg/mL (252.67 mM; Need ultrasonic and warming)
SMILES CC(C)C[C@@H](C(OC)=O)NC(C1=CC=C(NC[C@@H](N)CS)C=C1C2=C3C=CC=CC3=CC=C2)=O.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 593.66
Formula Weight 593.66
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.