missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GGT5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530042
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Mouse (81%), Chimpanzee (99%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of a gene family that encodes gamma-glutamyl transpeptidase enzymes. This enzyme consists of a heavy and a light chain, and is able to hydrolyze the gamma-glutamyl moiety of glutathione. It converts leukotriene C4 to leukotriene D4, however, it doesn't convert synthetic substrates that are commonly used to assay gamma-glutamyl transpeptidase. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| GGT5 | |
| Polyclonal | |
| Unconjugated | |
| GGT5 | |
| AI551243; gamma-glutamyl cleaving enzyme; gamma-glutamyl leukotrienase; Gamma-glutamyl transpeptidase-related enzyme; gamma-glutamyl transpeptidase-related protein; gamma-glutamyltransferase 5; Gamma-glutamyltransferase-like activity 1; gamma-glutamyltranspeptidase 5; GGL; GGT 5; GGT5; Ggta1; GGTLA1; GGT-rel; glutathione hydrolase 5; Glutathione hydrolase 5 heavy chain; Glutathione hydrolase 5 light chain; Glutathione hydrolase 5 proenzyme; Leukotriene-C4 hydrolase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2687 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.61 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P36269 | |
| GGT5 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human GGT5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction