missing translation for 'onlineSavingsMsg'
Learn More

GGT5 Antibody [CoraFluor™ 1], Novus Biologicals™

Product Code. 30732902 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732902 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732902 Supplier Novus Biologicals Supplier No. AF7209CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

GGT5 Polyclonal antibody specifically detects GGT5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGT5
Applications Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias DKFZp566O011, EC 2.3.2.2, gamma-glutamyl cleaving enzyme, Gamma-glutamyl transpeptidase-related enzyme, gamma-glutamyl transpeptidase-related protein, gamma-glutamyltransferase 5, Gamma-glutamyltransferase-like activity 1FLJ92733, Gamma-glutamyltranspeptidase 5, GGT 5, GGTLA1, GGT-rel
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GGT5, Ser30-Tyr586, Accession # P36269
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2687
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.