missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GGT2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA561540

Product Code. 15749351

  • £466.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: AVVQAIVRTA GGWAAASDSR KGGEPAGY Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 93%.

Isoform 1, isoform 2 and isoform 3 lack catalytic activity due to its inability to undergo the autocatalytic cleavage needed to produce a mature, enzymatically active heterodimer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GGT2
Polyclonal
Unconjugated
GGT2
gamma-glutamyltransferase 2; gamma-glutamyltranspeptidase 2; GGT; GGT 2; GGT2; glutathione hydrolase 2; inactive gamma-glutamyltranspeptidase 2
Rabbit
Antigen affinity chromatography
RUO
728441
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P36268
GGT2
Recombinant protein corresponding to Human GGT2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.