missing translation for 'onlineSavingsMsg'
Learn More

GGT1, Mouse anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16178644
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16178644 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16178644 Supplier Abnova Supplier No. H00002678B01P.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Sequence: MKKKLVVLGLLAVVLVLVIVGLCLWLPSASKEPDNHVYTRAAVAADAKQCSKIGRDALRDGGSAVDAAIAALLCVGLMNAHSMGIGGGLFLTIYNSTTRKAEVINAREVAPRLAFATMFNSSEQSQKGGLSVAVPGEIRGYELAHQRHGRLPWARLFQPSIQLARQGFPVGKGLAAALENKRTVIEQQPVLCEVFCRDRKVLREGERLTLPQLADTYETLAIEGAQAFYNGSLTAQIVKDIQAAGGIVTAEDLNNYRAELIEHPLNISLGDVVLYMPSAPLSGPVLALILNILKGYNFSRESVESPEQKGLTYHRIVEAFRFAYAKRTLLGDPKFVDVTEVVRNMTSEFFAAQLRAQISDDTTHPISYYKPEFYTPDDGGTAHLSVVAEDGSAVSATSTINLYFGSKVRSPVSGILFNNEMDDFSSPSITNEFGVPPSPANFIQPGKQPLSSMCPTIMVGQDGQVRMVVGAAGGTQITTATALAIIYNLWFGYDVKRAVEEPRLHNQLLPNVTTVERNIDQAVTAALETRHHHTQIASTFIAVVQAIVRTAGGWAAASDSRKGGEPAGY

Specifications

Antigen GGT1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene gamma-glutamyltransferase 1
Gene Accession No. NM_001032364.1
Gene Alias CD224/D22S672/D22S732/GGT/GTG/MGC96892/MGC96904/MGC96963
Gene Symbols GGT1
Host Species Mouse
Immunogen GGT1 (NP_005256.2, 1 a.a. ∼ 569 a.a) full-length human protein.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2678
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.