missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGPS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97048-50ul
This item is not returnable.
View return policy
Description
GGPS1 Polyclonal antibody specifically detects GGPS1 in Human samples. It is validated for ELISA
Specifications
| GGPS1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| 6E)-farnesyl diphosphate synthase, Dimethylallyltranstransferase, EC 2.5.1.1, EC 2.5.1.10, EC 2.5.1.29, Farnesyl diphosphate synthase, Farnesyltranstransferase, Geranylgeranyl diphosphate synthase, geranylgeranyl diphosphate synthase 1, geranylgeranyl pyrophosphate synthase, geranyltranstransferase, GGPP synthase, GGPPS, GGPPS1farnesyltranstransferase, GGPPSase | |
| Produced in rabbits immunized with purified, recombinant Human GGPS1 (Accession#: NP_001032354.1; Met1-Glu300) | |
| 50 μg | |
| Cancer | |
| 9453 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction