missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GGNBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553879
This item is not returnable.
View return policy
Description
Immunogen sequence: VLRAYNILIG ELDCSKEKGY CAALYEGLRC CPHERHIHVC CETDFIAHLL GRAEPEFAGG RRERHAKTID IAQEEVLTCL GIHLYERLHR IWQKLRAE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
May be involved in spermatogenesis. Its mouse homolog is induced by dioxin and may play a role in spermatogenesis.
Specifications
| GGNBP2 | |
| Polyclonal | |
| Unconjugated | |
| GGNBP2 | |
| AI451896; C3HC4-type zinc finger protein; C75939; D330017P12Rik; DIF3; DIF-3; dioxin inducible factor 3; dioxin-inducible factor 3; gametogenetin binding protein 2; gametogenetin-binding protein 2; gametogenin binding protein 2; Ggnbp2; laryngeal carcinoma related gene 1; laryngeal carcinoma-related protein 1; LCRG1; LZK1; Protein ZNF403; Zfp403; zinc finger protein 403; ZNF403 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79893 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H3C7 | |
| GGNBP2 | |
| Recombinant protein corresponding to Human GGNBP2. Recombinant protein control fragment (Product #RP-92273). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction