missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GGA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566608
This item is not returnable.
View return policy
Description
Immunogen sequence: NCCEEKRNPSS STLPGGGVQN PSADRNLLDL LSPQPAPCPL NYVSQKSVPK EVPPGTKSSP GWSWEAGPLA PSPSSQNTP Highest antigen sequence identity to the following orthologs - mouse 73%, rat 75%.
This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. This family member may play a significant role in cargo molecules regulation and clathrin-coated vesicle assembly.
Specifications
| GGA2 | |
| Polyclonal | |
| Unconjugated | |
| GGA2 | |
| 1200007E24Rik; ADP-ribosylation factor binding protein 2; ADP-ribosylation factor-binding protein GGA2; gam; Gamma-adaptin-related protein 2; Gga2; golgi associated, gamma adaptin ear containing, ARF binding protein 2; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; golgi-localized, gamma ear-containing, ARF-binding protein 2; Kiaa1080; mKIAA1080; VEAR; VHS domain and ear domain of gamma-adaptin; VHS domain and ear domain-containing protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23062 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UJY4 | |
| GGA2 | |
| Recombinant Human GGA2. Recombinant protein control fragment (Product #RP-107267). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction