missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GFR alpha-like Antibody (2345C) [PE/Atto594], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ FAB9697PEATT594
This item is not returnable.
View return policy
Description
GFR alpha-like Monoclonal antibody specifically detects GFR alpha-like in Human samples. It is validated for Flow Cytometry
Specifications
| GFR alpha-like | |
| Monoclonal | |
| PE/ATTO 594 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2345C | |
| PBS | |
| Human embryonic kidney cell line HEK293-derived recombinant human GFR alpha-like, Ser19-Glu351, Accession # Q6UXV0 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction