missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GFR alpha-3/GDNF R alpha-3 Antibody (nadecnemab) - IgG4SP, Novus Biologicals™

Human Monoclonal Antibody

£162.00 - £695.00

Specifications

Antigen GFR alpha-3/GDNF R alpha-3
Clone nadecnemab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30047643
View Documents
Novus Biologicals™
NBP3-28758-1mg
1 mg
£695.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30045082
View Documents
Novus Biologicals™
NBP3-28758-100ug
100 μg
£247.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30046741
View Documents
Novus Biologicals™
NBP3-28758-50ug
50 μg
£162.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GFR alpha-3/GDNF R alpha-3 Monoclonal antibody specifically detects GFR alpha-3/GDNF R alpha-3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

GFR alpha-3/GDNF R alpha-3
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Cytokine Research, Immunology
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
GDNF family receptor alpha 3, GDNF family receptor alpha-3, GDNF receptor alpha-3, GDNFR-alpha-3, GFRa-3, GFR-alpha-3, glial cell line-derived neurotrophic factor receptor alpha-3, GPI-linked receptor
GFRA3
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
nadecnemab
Monoclonal
Purified
RUO
Human
O60609
2676
IgG4
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.