missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFR alpha-2/GDNF R alpha-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | GFR alpha-2/GDNF R alpha-2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
GFR alpha-2/GDNF R alpha-2 Polyclonal specifically detects GFR alpha-2/GDNF R alpha-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| GFR alpha-2/GDNF R alpha-2 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Cytokine Research, Immunology | |
| PBS with 0.02% Sodium Azide | |
| GDNF family receptor alpha 2, GDNF family receptor alpha-2, GDNF receptor alpha-2, GDNF receptor beta, GDNFR-alpha-2, GDNFR-beta, GDNFRBRET ligand 2, GFR-alpha 2, GFR-alpha-2, glial cell line derived neurotrophic factor receptor, beta, Neurturin receptor alpha, NRTNR-alpha, NTNRA, NTNR-alpha, PI-linked cell-surface accessory protein, RETL2TGF-beta-related neurotrophic factor receptor 2, TGF-beta related neurotrophic factor receptor 2, TRN receptor, GPI-anchored, TRNR2NRTNR-ALPHA | |
| GFRA2 | |
| IgG | |
| Ion exchange chromatography | |
| 52 kDa |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| AAC52036 | |
| 2675 | |
| Antibody was raised against a 15 amino acid peptide near the center of human GFR alpha 2. The immunogen is located within amino acids 360 - 410 of GFR alpha 2. Amino Acid Squence: RVEKTPSLPDDLSDS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title