missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFR alpha-2/GDNF R alpha-2 Antibody (71515) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6131JF585
This item is not returnable.
View return policy
Description
GFR alpha-2/GDNF R alpha-2 Monoclonal antibody specifically detects GFR alpha-2/GDNF R alpha-2 in Human samples. It is validated for Immunocytochemistry
Specifications
| GFR alpha-2/GDNF R alpha-2 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| GDNF family receptor alpha 2, GDNF family receptor alpha-2, GDNF receptor alpha-2, GDNF receptor beta, GDNFR-alpha-2, GDNFR-beta, GDNFRBRET ligand 2, GFR-alpha 2, GFR-alpha-2, glial cell line derived neurotrophic factor receptor, beta, Neurturin receptor alpha, NRTNR-alpha, NTNRA, NTNR-alpha, PI-linked cell-surface accessory protein, RETL2TGF-beta-related neurotrophic factor receptor 2, TGF-beta related neurotrophic factor receptor 2, TRN receptor, GPI-anchored, TRNR2NRTNR-ALPHA | |
| Mouse myeloma cell line NS0-derived recombinant human GFR alpha -2/GDNF R alpha -2, Ser22-Ser441, Accession # O00451 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunocytochemistry | |
| 71515 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cytokine Research, Immunology | |
| 2675 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction