missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFPT1 Recombinant Rabbit Monoclonal Antibody (K01_1R64)
GREENER_CHOICE

Product Code. 30830103
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830103 100 μL
30830322 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830103 Supplier Invitrogen™ Supplier No. MA563707

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

GFPT1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes the first and rate-limiting enzyme of the hexosamine pathway and controls the flux of glucose into the hexosamine pathway. The product of this gene catalyzes the formation of glucosamine 6-phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFPT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1R64
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene GFPT1
Gene Accession No. Q06210
Gene Alias 2810423A18Rik; AI324119; AI449986; CMS12; CMSTA1; D-fructose-6-phosphate amidotransferase 1; eartha; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; Gfpt; GFPT1; GFPT1L; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine fructose-6-phosphate transaminase 1; glutamine:fructose 6 phosphate amidotransferase 1; glutamine:fructose-6-phosphate amidotransferase 1; glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine--fructose-6-phosphate transaminase 1; harvest moon; hexosephosphate aminotransferase 1; hmn; MSLG; unm_z110
Gene Symbols GFPT1
Host Species Rabbit
Immunogen A synthetic peptide of human GFPT1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2673
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.