missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFPT1 Recombinant Rabbit Monoclonal Antibody (24GB3030)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557032

 View more versions of this product

Product Code. 30274799

  • £126.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GFPT1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

This gene encodes the first and rate-limiting enzyme of the hexosamine pathway and controls the flux of glucose into the hexosamine pathway. The product of this gene catalyzes the formation of glucosamine 6-phosphate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GFPT1
Recombinant Monoclonal
3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P47856, P82808, Q06210
GFPT1
A synthesized peptide derived from human GFPT1 (aa 620-660).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot, Immunocytochemistry
24GB3030
Unconjugated
GFPT1
2810423A18Rik; AI324119; AI449986; CMS12; CMSTA1; D-fructose-6-phosphate amidotransferase 1; eartha; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; Gfpt; GFPT1; GFPT1L; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine fructose-6-phosphate transaminase 1; glutamine:fructose 6 phosphate amidotransferase 1; glutamine:fructose-6-phosphate amidotransferase 1; glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine--fructose-6-phosphate transaminase 1; harvest moon; hexosephosphate aminotransferase 1; hmn; MSLG; unm_z110
Rabbit
Affinity Chromatography
RUO
14583, 2673, 297417
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.