missing translation for 'onlineSavingsMsg'
Learn More

GFP-STAT1

Product Code. 10637805
Click to view available options
Quantity:
10 nmol
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10637805

Brand: Invitrogen PV5211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

GFP-STAT1

STAT1 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for the Janus family of tyrosine kinases.

For use in combination with the LanthaScreen™ Tb-anti-STAT1 [pTyr701] Antibody (PV4844) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Biochemical terbium phospho-specific activity
Content And Storage GFP-STAT1 protein substrate in 50 mM Tris-HCl pH 7.5, 100 mM NaCl, 10% Glycerol, 1 mM DTT. Store at -80°C.
Detection Method Fluorescence
Quantity 10 nmol
Substrate STAT1
Substrate Properties Protein-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Tyrosine Kinases
Label or Dye GFP (EmGFP)
Molecular Weight (g/mol) 116000 Da
Shipping Condition Dry Ice
Target Entry JAK1, JAK2 JH1 JH2, JAK2, TYK2, JAK3, JAK2 JH1 JH2 V617F
Validated Kinases JAK1, JAK2, JAK3, TYK2
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.