Learn More
GFP Monoclonal Antibody (5F12.4), eFluor™ 660, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 50-6498-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: This 5F12.4 monoclonal antibody reacts with green-fluorescent protein (GFP), which was originally isolated from the cnidarian Aequorea victoria. This protein absorbs blue light (maximally at 395 nm) and emits green light (peak at 509) without the requirement of exogenous substrates and cofactors. These unique qualities allow GFP to be used to monitor gene expression and protein localization in vivo. Several mutant forms of GFP have been developed which fluoresce more intensely and have shifted excitation maxima when compared to wildtype GFP, making them useful for flow cytometry, fluorescence microscopy, and double-labeling applications. This antibody is capable of staining formaldehyde fixed cells. Applications Reported: This 5F12.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 5F12.4 antibody has been tested by flow cytometric analysis of GFP expressing cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm).
GFP Monoclonal antibody specifically detects GFP in Tag samples. It is validated for Flow Cytometry.Specifications
GFP | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
100 μg | |
Primary | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
5F12.4 | |
eFluor 660 | |
GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP | |
Affinity chromatography | |
RUO | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid | |
Tag |
For Research Use Only.