missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFP Polyclonal Antibody, Alexa Fluor™ 488

Product Code. 10797644
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10797644 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10797644 Supplier Invitrogen™ Supplier No. A21311
  • ONLINE EXCLUSIVE PRICE
    £488.00
    £439.65 / 100µL
    Save £48.35
    9% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

To achieve appropriate staining results on formalin-fixed, paraffin embedded tissue sections, protease Type XIV (Actinase E) pretreatment is required. The recommended dilution and incubation for protease is 1-2 mg/mL at 37°C for 10 minutes. Other enzyme and Heat Induced Epitope Retrieval pre-treatment methods may not be suitable for this particular antibody. Frozen sections have been successfully stained after fixation with acetone at 4°C.

The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFP
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 5mM sodium azide; pH 7.5
Gene Alias GFP; GFP tag; GFP2; green fluorescence; green fluorescent; green fluorescent protein; Turbo eGFP
Host Species Rabbit
Immunogen The GFP was isolated directly from the jellyfish Aequorea victoria.
Purification Method IgG fraction
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.