missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Mouse, DyLight 594, Clone: 1025527, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB42402M
This item is not returnable.
View return policy
Description
GFP Monoclonal antibody specifically detects GFP in Broad Species Reactivity samples. It is validated for Western Blot, Flow Cytometry
Specifications
| GFP | |
| Monoclonal | |
| DyLight 594 | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| E. coli-derived recombinant GFP, aa 2-238 | |
| 0.1 mL | |
| Epitope Tags | |
| All species | |
| Purified |
| Western Blot, Flow Cytometry | |
| 1025527 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction