missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Monoclonal Antibody (GF28R), Biotin, Invitrogen™
Description
MA5-15256-BTIN has been successfully used for Western blot, ELISA, ICC/IF and IP applications.
GFP Monoclonal antibody specifically detects GFP in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.
Specifications
Specifications
| Antigen | GFP |
| Applications | ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | GF28R |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with proprietary stabilizer and 0.02% sodium azide |
| Gene Alias | GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP |
| Host Species | Mouse |
| Immunogen | GFP from the jellyfish Aequorea Victoria N-terminal peptide-KLH conjugated. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?