Learn More
Invitrogen™ GFP Monoclonal Antibody (3E6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ A11120
294.75 GBP valid until 2025-12-19
BEST PRICE on Promo! Use promo code "24110" to get your promotional price.
Description
Storage and reconstitution: to prepare a 0.2 mg/mL stock solution, reconstitute the lyophilized antibodies in 0.5mL PBS, pH 7.4. Store solution for up to 3months at 4°C with the addition of 2mM sodium azide.
The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
Specifications
| GFP | |
| Monoclonal | |
| Unconjugated | |
| GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP | |
| The GFP was isolated directly from the jellyfish Aequorea victoria. | |
| 100 μg | |
| Primary | |
| -20°C | |
| Lyophilized | |
| Tag |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 3E6 | |
| PBS with BSA and no preservative | |
| Mouse | |
| Protein G | |
| RUO | |
| Tag | |
| Antibody | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.