missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GFP Monoclonal Antibody (3B11)
Description
GFP Monoclonal Antibody for Western Blot, ICC/IF, IHC
The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
Specifications
Specifications
| Antigen | GFP |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3B11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 5mM sodium azide |
| Gene Alias | GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP |
| Host Species | Mouse |
| Immunogen | Recombinant protein purified from E. coli. The epitope is in the N-terminal 18 amino acids of the protein, the peptide MVSKGAELFTGIVPILIE, which is found in the Clontech and other GFP vectors. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?