missing translation for 'onlineSavingsMsg'
Learn More

GFP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£210.00 - £977.00

Specifications

Target Species Non-species specific, C. elegans, Plant
Gene Symbols GFP
Purification Method Affinity Purified
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18750264
View Documents
Novus Biologicals
NB600-308SS
£210.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18730254
View Documents
Novus Biologicals
NB600-308
£410.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30156535
View Documents
Novus Biologicals
NB600-308-1mg
1 mg
£977.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Electron Microscopy, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockdown Validated.
Specifications

Specifications

Non-species specific, C. elegans, Plant
GFP
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.