missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-56401SS
This item is not returnable.
View return policy
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot.
Specifications
| GFP | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Non-species specific | |
| Purified |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 0.5-2 ug/ml | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| A synthetic peptide corresponding to amino acids 102-118 (KDDGNYKTRAEVKFEGD) of GFP were used as immunogen for this antibody. | |
| 0.025 mg | |
| Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
| The GFP amino acid sequence used as immunogen for this antibody is 100% homologous in YFP (Yellow) and CFP (Cyan). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction