Learn More
GFP Antibody (4B10B2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-80750
Product Code. 18295978
  • £401.00 / 0.10mL
Description
GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GFP
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000. Use reported by customer review, Flow Cytometry 1:200-1:400, ELISA : 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence : 1:10-1:500, Immunohistochemistry-Paraffin : 1:200-1:1000, CyTOF-ready
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
This GFP antibody was developed against purified recombinant fragment of GFP expressed in E. coli.
0.1 mL
Primary
Non-species specific
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
4B10B2
Unconjugated
PBS
Mouse
Ammonium sulfate precipitation
RUO
Reacts with GFP.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a