missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GFM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111521
This item is not returnable.
View return policy
Description
Immunogen sequence: NKLDRMGSNP ARALQQMRSK LNHNAAFMQI PMGLEGNFKG IVDLIEERAI YFDGDFGQIV RYGEIPAELR AAATDHRQEL IECVANSDEQ LGEMFLE Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 92%.
Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. Mitochondrial translation is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. This gene encodes one of the mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known.
Specifications
| GFM1 | |
| Polyclonal | |
| Unconjugated | |
| GFM1 | |
| AW545374; COXPD1; D3Wsu133e; Efg; EF-G; Efg1; EFGM; EF-Gmt; EF-Gmt {ECO:0000255; EGF1; Elongation factor G 1, mitochondrial; elongation factor G 1, mitochondrial {ECO:0000255; elongation factor G, mitochondrial; elongation factor G, mitochondrial {ECO:0000255; Elongation factor G1; elongation factor G1 {ECO:0000255; G elongation factor 1; G elongation factor mitochondrial 1; G elongation factor, mitochondrial 1; G translation elongation factor, mitochondrial; GFM; Gfm1; HAMAP-Rule:MF_03061}; hEFG1; mEF-G 1; mEF-G 1 {ECO:0000255; mitochondrial; mitochondrial elongation factor G; mitochondrial elongation factor G1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85476 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96RP9 | |
| GFM1 | |
| Recombinant Protein corresonding to Human GFM1. Recombinant protein control fragment (Product #RP-105185). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction