missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GFI1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540328
This item is not returnable.
View return policy
Description
Peptide sequence: MPRSFLVKSK MAHTYHQPRV QEDEPLWPPA LTPVPRDQAP SNSPVLSTLF Sequence homology: Cow: 83%; Dog: 100%; Goat: 83%; Horse: 100%; Human: 100%; Mouse: 83%; Pig: 93%; Rat: 82%.
GFI1b (zinc finger protein GFI-1b) is an essential proto-oncogenic transcriptional regulator necessary for development and differentiation of erythroid and megakaryocytic lineages. It is also a component of a RCOR-GFI-KDM1A-HDAC complex that suppresses, via histone deacetylase (HDAC) recruitment, a number of genes implicated in multilineage blood cell development and controls hematopoietic differentiation. GFI1b also acts as a transcriptional repressor or activator depending on both promoter and cell type context; represses promoter activity of SOCS1 and SOCS3 and thus, may regulate cytokine signaling pathways. It also cooperates with GATA1 to repress target gene transcription, such as the apoptosis regulator BCL2L1; GFI1B silencing in leukemic cell lines markedly increase apoptosis rate. Mutations affecting the gene are associated with Bleeding disorder, platelet-type 17 (BDPLT17).
Specifications
| GFI1B | |
| Polyclonal | |
| Unconjugated | |
| GFI1B | |
| BDPLT17; GFI1B; Gfi-1B; growth factor independent 1B; growth factor independent 1B (potential regulator of CDKN1A, translocated in CML); growth factor independent 1B transcription repressor; growth factor independent 1B transcriptional repressor; growth factor independent 1B-like protein; growth factor independent protein 1B; growth factor-independent 1B; Potential regulator of CDKN1A translocated in CML; Zinc finger protein Gfi-1b; ZNF163B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8328 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5VTD9 | |
| GFI1B | |
| Synthetic peptide directed towards the N-terminal of human GFI1B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction