missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GFI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577985
This item is not returnable.
View return policy
Description
Positive Control: THP-1, HL-60 Predicted Reactivity: Mouse (80%), Rat (81%), Dog (87%), Cat (89%), Rhesus Monkey (98%), Bovine (88%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GFI1 is a nuclear zinc finger (ZF) transcriptional repressor that plays an important role in hematopoiesis and inner ear development, and has been implicated in lymphomagenesis. The Gfi1 protein expression is regulated by the ubiquitin-proteasome system. It is reported that ubiquitin ligase Triad1 interacts with the DNA-binding domain of Gfi-1. Gfi-1 represses transcription by directly binding to the consensus DNA sequence in the promoters of its target genes. Deficiency of Gfi-1 causes neutropenia and an accumulation of granulomonocytic precursor cells that is reminiscent of a myelodysplastic syndrome in mice.
Specifications
| GFI1 | |
| Polyclonal | |
| Unconjugated | |
| GFI1 | |
| AW495828; FLJ94509; Gfi1; GFI-1; GFI1A; growth factor independence-1; growth factor independent 1; growth factor independent 1 transcription repressor; growth factor independent 1 transcriptional repressor; growth factor independent protein 1; Growth factor independent-1; Pal1; Pal-1; SCN2; Zinc finger protein 163; zinc finger protein Gfi-1; ZNF163 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2672 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.49 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q99684 | |
| GFI1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human GFI1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering