missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFI-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£158.00 - £360.00
Specifications
| Antigen | GFI-1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 6-8ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18701284
|
Novus Biologicals
NBP2-27173SS |
0.025 mg |
£158.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18739973
|
Novus Biologicals
NBP2-27173 |
0.1 mg |
£360.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFI-1 Polyclonal specifically detects GFI-1 in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GFI-1 | |
| Western Blot 6-8ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine, Chicken | |
| GFI-1, growth factor independence-1, growth factor independent 1, growth factor independent 1 transcription repressor, Growth factor independent protein 1, SCN2, Zinc finger protein 163, zinc finger protein Gfi-1, ZNF163FLJ94509 | |
| GFI1 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chicken and cow. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 2672 | |
| A portion of amino acid 280-330 of human Zinc finger protein Gfi-1 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts