missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFER Monoclonal Antibody (829812)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561889

Product Code. 30534899

  • £315.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

Isoform 1: FAD-dependent sulfhydryl oxidase that regenerates the redox-active disulfide bonds in CHCHD4/MIA40, a chaperone essential for disulfide bond formation and protein folding in the mitochondrial intermembrane space. The reduced form of CHCHD4/MIA40 forms a transient intermolecular disulfide bridge with GFER/ERV1, resulting in regeneration of the essential disulfide bonds in CHCHD4/MIA40, while GFER/ERV1 becomes re- oxidized by donating electrons to cytochrome c or molecular oxygen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GFER
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P55789
Gfer
E. coli-derived recombinant human GFER/ALR, Arg82-Asp205.
100 μg
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
829812
Unconjugated
Gfer
ALR; Augmenter of liver regeneration; ERV1; ERV1 homolog; erv1-like growth factor; FAD-linked sulfhydryl oxidase ALR; Gfer; growth factor, augmenter of liver regeneration; growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration); growth factor, erv1 homolog; growth factor, erv1-like (augmenter of liver regeneration); hepatic regenerative stimulation substance; Hepatopoietin; hepatopoietin protein; HERV1; HPO; HPO1; HPO2; HSS; MNCb-0663
Mouse
Protein A/G
RUO
2671
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.