missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFER/ALR Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97019CL1
This item is not returnable.
View return policy
Description
GFER/ALR Polyclonal antibody specifically detects GFER/ALR in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| GFER/ALR | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| ALRFAD-linked sulfhydryl oxidase ALR, Augmenter of liver regeneration, EC 1.8.3.2, ERV1 homolog, ERV1hepatopoietin protein, erv1-like growth factor, growth factor, augmenter of liver regeneration, growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration), Hepatopoietin, hERV1, HERV1hepatic regenerative stimulation substance, HPO, HPO1, HPO2, HSS, truncated augmenter of liver regeneration | |
| Produced in rabbits immunized with purified, recombinant Human GFER/ALR (Accession#: NP_005253.3; Met1-Asp125) | |
| 0.1 mL | |
| Cell Biology, Endocrinology, Signal Transduction | |
| 2671 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction