missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Mouse, Clone: SPM248, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-34353-0.1MG
This item is not returnable.
View return policy
Description
GFAP Monoclonal specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GFAP | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| FLJ45472, GFAP astrocytes, glial fibrillary acidic protein | |
| Mouse | |
| 50 kDa | |
| 0.1 mg | |
| Astrocyte Markers, Cancer, Cellular Markers, Cytoskeleton Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| 2670 | |
| Human, Mouse, Rat, Pig, Bovine, Chicken, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SPM248 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
| P14136 | |
| GFAP | |
| GFAP isolated from pig spinal cord | |
| Protein G purified | |
| RUO | |
| Primary | |
| This MAb recognizes a protein of ∼50kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins. GFAP is specifically found in astroglia. GFAP is a very popular marker for localizing benign astrocyte and neoplastic cells of glial origin in the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS. | |
| Store at 4C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction