missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4B2E10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GFAP |
|---|---|
| Clone | 4B2E10 |
| Concentration | 1.5 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16887175
|
Proteintech
60190-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16877175
|
Proteintech
60190-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFAP (Glial fibrillary acidic protein) is a member of the class III intermediate filament protein family. GFAP is heavily, and specifically, expressed in astrocytes and certain astroglia in the central nervous system, in satellite cells in peripheral ganglia, and in non-myelinating Schwann cells in peripheral nerves. In addition, neural stem cells strongly express GFAP. Antibodies to GFAP are very useful as markers of astrocytic cells. In addition, many types of brain tumor, presumably derived from astrocytic cells, heavily express GFAP. GFAP is also found in the lens epithelium, Kupffer cells of the liver, in some cells in salivary tumors and has been reported in erythrocytes. GFAP is used as a marker to distinguish astrocytes from other glial cells during development. Mutations in this gene cause Alexander disease, a rare disorder of astrocytes in the central nervous system. Alternative splicing of the GFAP gene results in multiple transcript variants encoding distinct isoforms.Specifications
| GFAP | |
| 1.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GFAP | |
| GFAP | |
| IgG2a | |
| Protein A | |
| Antibody |
| 4B2E10 | |
| Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Rat, Mouse, Human, Pig | |
| P03995, P14136, P47819 | |
| 14580, 24387, 2670, 396562 | |
| GFAP Fusion Protein Ag10452 | |
| Primary | |
| -20°C | |
| GFAP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title