missing translation for 'onlineSavingsMsg'
Learn More

GFAP (Glial Fibrillary Acidic Protein) Mouse Monoclonal Antibody, Epredia™

Product Code. 12659686 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
Unit Size:
100µL
1mL
200µL
500µL
7mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form Concentration unitSize
12659686 500 μL Concentrated 200μg/mL 500µL
12649686 100 μL Concentrated 200μg/mL 100µL
12679686 100 μL Concentrated 1mg/mL 100µL
12669686 200 μL Concentrated 1mg/mL 200µL
12639686 1 mL Concentrated 200μg/mL 1mL
12619696 7 mL Prediluted Prediluted 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12659686 Supplier Epredia Supplier No. MS1376P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Glial Fibrillary Acidic Protein (GFAP) is specific to astrocytes (i.e., glial cells) and ependymal cells of the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS. Ab-6 stains astrocytes, glial cells, ependymal cells and their corresponding tumors. Many types of neural tumors such as neuroblastomas, Schwannomas, as well as extra-CNS tumors are not labeled.

Host Species: Mouse

Clone: ASTRO6

Isotype: IgG1

Species Reactivity: Human, Pig, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Glial Fibrillary Acidic Protein

Molecular Weight: 51-52 kDa

Positive Control: IMR-5 cells, Brain or astrocytoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFAP (Glial Fibrillary Acidic Protein)
Applications Immunofluorescence
Classification Monoclonal
Clone ASTRO6
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Glial Fibrillary Acidic Protein
Quantity 500 μL
Regulatory Status IVD
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Chicken, Human, Pig, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.