missing translation for 'onlineSavingsMsg'
Learn More

GFAP Antibody (5C10), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18430297 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430297 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430297 Supplier Novus Biologicals Supplier No. NBP105197MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Free-Floating
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFAP
Applications Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 5C10
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry - Free Floating
Formulation 50mM Sodium Borate
Gene Alias ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
Host Species Mouse
Immunogen This GFAP Antibody (5C10) was developed against a preparation of purified pig spinal cord GFAP
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2670
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.